Learn More
Biomatik Corporation Mouse Nicotinamide-N-Methyltransferase (NNMT) ELISA Kit
Supplier:  Biomatik Corporation EKU06297
Encompass_Preferred
Mouse Nicotinamide-N-Methyltransferase (NNMT) ELISA Kit for detecting NNMT in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O55239. Gene ID: 18113. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2727