Learn More
Biomatik Corporation Pig Endothelial NOS (eNOS) ELISA Kit
Supplier:  Biomatik Corporation EKU06314
Encompass_Preferred
Pig Endothelial NOS (eNOS) ELISA Kit for detecting eNOS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.59ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28969. Gene ID: 397557. Target Synonyms: NOS3; ECNOS; NOS III; E-NOS; C-NOS; ENOS; CNOS; Constitutive NOS; Nitric Oxide Synthase 3, Endothelial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2729