Learn More
Biomatik Corporation Rat Nuclear Factor I/X (NFIX) ELISA Kit
Supplier:  Biomatik Corporation EKU06350
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: F2Z3R4. Target Synonyms: NF1A; CTF; CCAAT-Binding Transcription Factor; TGGCA-binding protein. Restriction: For Research Use Only.
Catalog No. 50-229-2733