Learn More
Biomatik Corporation Rat Glucocorticoid Receptor (GR) ELISA Kit
Supplier:  Biomatik Corporation EKU06368
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P06536. Target Synonyms: GCCR; GCR; NR3C1; GRL; NR3-C1; Nuclear Receptor Subfamily 3, Group C, Member 1. Restriction: For Research Use Only.
Catalog No. 50-229-2735