Learn More
Biomatik Corporation Human Nucleoporin 62 (NUP62) ELISA Kit
Supplier:  Biomatik Corporation EKU06376
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P37198. Target Synonyms: IBSN; p62; SNDI; Nuclear Pore Glycoprotein p62; Nucleoporin 62kDa. Restriction: For Research Use Only.
Catalog No. 50-229-2737