Learn More
Biomatik Corporation Mouse Palate/Lung And Nasal Epithelium Associated Protein (PLUNC) ELISA Kit
Supplier:  Biomatik Corporation EKU06463
Encompass_Preferred
Mouse Palate/Lung And Nasal Epithelium Associated Protein (PLUNC) ELISA Kit for detecting PLUNC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97361. Gene ID: 18843. Target Synonyms: BPIFA1; SPURT; NASG; LUNX; SPLUNC1; Secretory Protein In Upper Respiratory Tracts; BPI Fold Containing Family A, Member 1; Nasopharyngeal carcinoma-related. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2750