Learn More
Biomatik Corporation Rat Perforin 1 (PRF1) ELISA Kit
Supplier:  Biomatik Corporation EKU06529
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P35763. Target Synonyms: FLH2; HPLH2; P1; PFP; Pore Forming Protein; Cytolysin; Lymphocyte pore-forming protein. Restriction: For Research Use Only.
Catalog No. 50-229-2757