Learn More
Biomatik Corporation Mouse Perilipin 4 (PLIN4) ELISA Kit
Supplier:  Biomatik Corporation EKU06534
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O88492. Target Synonyms: Adipocyte protein S3-12. Restriction: For Research Use Only.
Catalog No. 50-229-2759