Learn More
Biomatik Corporation Human Peroxiredoxin 4 (PRDX4) ELISA Kit
Supplier:  Biomatik Corporation EKU06554
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Human. Detection Range: 62.5-4,000pg/mL. Sensitivity: 26.7pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q13162. Target Synonyms: Antioxidant enzyme AOE372; Thioredoxin-dependent peroxide reductase A0372. Restriction: For Research Use Only.
Catalog No. 50-229-2761