Learn More
Biomatik Corporation Human Phospholipase A2, Group XIIB (PLA2G12B) ELISA Kit
Supplier:  Biomatik Corporation EKU06622
Encompass_Preferred
Human Phospholipase A2, Group XIIB (PLA2G12B) ELISA Kit for detecting PLA2G12B in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BX93. Gene ID: 84647. Target Synonyms: PLA2G13; FKSG71; GXIIB; Phospholipase A2,Group XIII; Group XIIB secretory phospholipase A2-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2768