Learn More
Biomatik Corporation Mouse Placenta Growth Factor (PLGF) ELISA Kit
Supplier:  Biomatik Corporation EKU06646
Encompass_Preferred
Mouse Placenta Growth Factor (PLGF) ELISA Kit for detecting PLGF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-500pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49764. Gene ID: 18654. Target Synonyms: PlGF2; PGF; PGFL; Placental Growth Factor-Like; Vascular Endothelial Growth Factor-Related Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2772