Learn More
Biomatik Corporation Pig Plasminogen (Plg) ELISA Kit
Supplier:  Biomatik Corporation EKU06656
Encompass_Preferred
Pig Plasminogen (Plg) ELISA Kit for detecting Plg in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000ng/ml. Sensitivity: 5.6ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06867. Gene ID: 733660. Target Synonyms: PL; Plasmin; Activation peptide; Angiostatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2774