Learn More
Biomatik Corporation Bovine Platelet Factor 4 (PF4) ELISA Kit
Supplier:  Biomatik Corporation EKU06702
Encompass_Preferred
Sample Type: Serum, platelet-poor plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Bovine. Detection Range: 6.25-400pg/mL. Sensitivity: 2.85pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P02777. Target Synonyms: CXCL4; SCYB4; Chemokine C-X-C-Motif Ligand 4; Oncostatin-A; Iroplact. Restriction: For Research Use Only.
Catalog No. 50-229-2780