Learn More
Biomatik Corporation Human Procollagen II (PCII) ELISA Kit
Supplier:  Biomatik Corporation EKU06776
Encompass_Preferred
Human Procollagen II (PCII) ELISA Kit for detecting PCII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: PCOL-II; PC-II; PC2; Type II Procollagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2790