Learn More
Biomatik Corporation Human Pro-Insulin Like Growth Factor II E-Peptide (IGFIIE) ELISA Kit
Supplier:  Biomatik Corporation EKU06812
Encompass_Preferred
Human Pro-Insulin Like Growth Factor II E-Peptide (IGFIIE) ELISA Kit for detecting IGFIIE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 37.0-3,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01344. Gene ID: 3481. Target Synonyms: Preptin; Proinsulin-Like Growth Factor-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2792