Learn More
Biomatik Corporation Rabbit Prolactin (PRL) ELISA Kit
Supplier:  Biomatik Corporation EKU06824
Encompass_Preferred
Rabbit Prolactin (PRL) ELISA Kit for detecting PRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.95ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q28632. Gene ID: 100009394. Target Synonyms: LTH; Luteotropic Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2795