Learn More
Biomatik Corporation Human Proteasome Subunit Beta Type 6 (PSMb6) ELISA Kit
Supplier:  Biomatik Corporation EKU06888
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.123ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P28072. Target Synonyms: LMPY; Macropain delta chain; Proteasome subunit Y; Multicatalytic endopeptidase complex delta chain. Restriction: For Research Use Only.
Catalog No. 50-229-2804