Learn More
Biomatik Corporation Human Protein Tyrosine Phosphatase, Non Receptor Type 6 (PTPN6) ELISA Kit
Supplier:  Biomatik Corporation EKU06951
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.121ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P29350. Target Synonyms: HCP; HCPH; HPTP1C; SH-PTP1; SHP-1L; SHP1; Hematopoietic cell protein-tyrosine phosphatase; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1. Restriction: For Research Use Only.
Catalog No. 50-229-2816