Learn More
Biomatik Corporation Rat Prothrombin Fragment 1+2 (F1+2) ELISA Kit
Supplier:  Biomatik Corporation EKU06967
Encompass_Preferred
Rat Prothrombin Fragment 1+2 (F1+2) ELISA Kit for detecting F1+2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18292. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2818