Learn More
Biomatik Corporation Human Pyruvate dehydrogenase alpha 1 (PDHA1) ELISA Kit
Supplier:  Biomatik Corporation EKU06985
Encompass_Preferred
Human Pyruvate dehydrogenase alpha 1 (PDHA1) ELISA Kit for detecting PDHA1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08559. Gene ID: 5160. Target Synonyms: PDHCE1A; PHE1A; PHE1-A; PDHa; PDH-A; Pyruvate Dehydrogenase (lipoamide) Alpha 1; Pyruvate Dehydrogenase E1 Component Subunit Alpha,Aomatic Form,Mitochondrial; Pyruvate Dehydrogenase Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2821