Learn More
Biomatik Corporation Human RAB1A, Member RAS Oncogene Family (RAB1A) ELISA Kit
Supplier:  Biomatik Corporation EKU07000
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.067ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P62820. Target Synonyms: RAB1; YPT1; Ras-related protein Rab-1A; YPT1-related protein. Restriction: For Research Use Only.
Catalog No. 50-229-2825