Learn More
Biomatik Corporation Rat RAR Related Orphan Receptor Gamma (RORg) ELISA Kit
Supplier:  Biomatik Corporation EKU07011
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: RORC; NR1F3; RORG; RZRG; TOR; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan receptor C; Retinoid-related orphan receptor-gamma. Restriction: For Research Use Only.
Catalog No. 50-229-2827