Learn More
Biomatik Corporation Human Cluster Of Differentiation 32 (CD32) ELISA Kit
Supplier:  Biomatik Corporation EKU07029
Encompass_Preferred
Human Cluster Of Differentiation 32 (CD32) ELISA Kit for detecting CD32 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.88-3,000pg/ml. Sensitivity: 17.35pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12318. Gene ID: 2212. Target Synonyms: FcgRII; FCGR2A; FCG2; FCGR2A1; FCGR2; FCGR2B; FCGR2C; Receptor II For The Fc Region Of Immunoglobulin G; Fc Fragment Of IgG, Low Affinity IIa, Receptor; Low affinity immunoglobulin gamma Fc region rec. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2831