Learn More
Biomatik Corporation Mouse Ryanodine Receptor 1, Skeletal (RYR1) ELISA Kit
Supplier:  Biomatik Corporation EKU07151
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.053ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CCO; MHS; MHS1; RYDR; RYR; SKRR; Central Core Disease Of Muscle; Skeletal muscle calcium release channel. Restriction: For Research Use Only.
Catalog No. 50-229-2851