Learn More
Biomatik Corporation Rat S100 Calcium Binding Protein A10 (S100A10) ELISA Kit
Supplier:  Biomatik Corporation EKU07160
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P05943. Target Synonyms: ANX2L; ANX2LG; CAL1L; CLP11; GP11; P11; P10; Calpactin-1 light chain; Cellular ligand of annexin II; Annexin II Ligand,Calpactin I,Light Polypeptide. Restriction: For Research Use Only.
Catalog No. 50-229-2853