Learn More
Biomatik Corporation Mouse Secreted Frizzled Related Protein 2 (SFRP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07217
Encompass_Preferred
Mouse Secreted Frizzled Related Protein 2 (SFRP2) ELISA Kit for detecting SFRP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 125-8,000pg/ml. Sensitivity: 46pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97299. Gene ID: 20319. Target Synonyms: FRP-2; SARP1; SDF-5; Secreted apoptosis-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2859