Learn More
Biomatik Corporation Rat Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07221
Encompass_Preferred
Rat Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit for detecting SFRP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JLS4. Gene ID: 89803. Target Synonyms: FRP-4; FRPHE; Frizzled protein, human endometrium. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2861