Learn More
Biomatik Corporation Human Semaphorin 3B (SEMA3B) ELISA Kit
Supplier:  Biomatik Corporation EKU07260
Encompass_Preferred
Human Semaphorin 3B (SEMA3B) ELISA Kit for detecting SEMA3B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13214. Gene ID: 7869. Target Synonyms: LUCA-1; SEMA5; SEMAA; SemA; semaV; Semaphorin-V; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2865