Learn More
Biomatik Corporation Mouse Sirtuin 3 (SIRT3) ELISA Kit
Supplier:  Biomatik Corporation EKU07353
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8R104. Target Synonyms: SIR2L3; Silent Mating Type Information Regulation 2 Homolog 3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; SIR2-like protein 3; Regulatory protein SIR2 homolog 3. Restriction: For Research Use Only.
Catalog No. 50-229-2881