Learn More
Biomatik Corporation Human Small Breast Epithelial Mucin (SBEM) ELISA Kit
Supplier:  Biomatik Corporation EKU07374
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.127ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q96DR8. Target Synonyms: MUCL1; Mucin-Like Protein 1; Protein BS106; Mucin-like protein 1. Restriction: For Research Use Only.
Catalog No. 50-229-2884