Learn More
Biomatik Corporation Mouse Solute Carrier Family 30, Member 3 (SLC30A3) ELISA Kit
Supplier:  Biomatik Corporation EKU07404
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P97441. Target Synonyms: ZNT3; Zinc Transporter 3. Restriction: For Research Use Only.
Catalog No. 50-229-2889