Learn More
Biomatik Corporation Human Sphingosine 1 Phosphate Receptor 3 (S1PR3) ELISA Kit
Supplier:  Biomatik Corporation EKU07435
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.60ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: EDG3; LPB3; S1P3; S1PR3; endothelial differentiation, sphingolipid G-protein-coupled receptor, 3; Sphingosine 1-phosphate receptor Edg-3. Restriction: For Research Use Only.
Catalog No. 50-229-2895