Learn More
Biomatik Corporation Human Suppression Of Tumorigenicity 14 (ST14) ELISA Kit
Supplier:  Biomatik Corporation EKU07511
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.60ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: AAH30532.1. Target Synonyms: SNC19; HAI; MT-SP1; PRSS14; TADG15; Epithin; Matriptase; Prostamin; Membrane-type serine protease 1; Serine protease 14; Tumor-associated differentially-expressed gene 15. Restriction: For Research Use Only.
Catalog No. 50-229-2908