Learn More
Biomatik Corporation Rat Surfactant Protein B (SP-B) ELISA Kit
Supplier:  Biomatik Corporation EKU07526
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, lung lavage fluid and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P22355. Target Synonyms: SFTPB; PSPB; SFTB3; SFTP3; SPB; Surfactant Associated Protein B; Pulmonary Surfactant Protein B; 18 kDa pulmonary-surfactant protein; 6 kDa protein. Restriction: For Research Use Only.
Catalog No. 50-229-2911