Learn More
Biomatik Corporation Human Tafazzin (TAZ) ELISA Kit
Supplier:  Biomatik Corporation EKU07565
Encompass_Preferred
Human Tafazzin (TAZ) ELISA Kit for detecting TAZ in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16635. Gene ID: 6901. Target Synonyms: CMD3A; EFE2; EFE; BTHS; XAP-2; G4.5; Barth syndrome; Endocardial Fibroelastosis 2; Cardiomyopathy,Dilated 3A (X-Linked). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2914