Learn More
Biomatik Corporation Human Thioredoxin Reductase 1 (TXNRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU07619
Encompass_Preferred
Human Thioredoxin Reductase 1 (TXNRD1) ELISA Kit for detecting TXNRD1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16881. Gene ID: 7296. Target Synonyms: TrxR1; TR1; GRIM12; KDRF; TRXR-1; TXNR; Gene associated with retinoic and interferon-induced mortality 12 protein; KM-102-derived reductase-like factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2926