Learn More
Biomatik Corporation Dog Thrombomodulin (TM) ELISA Kit
Supplier:  Biomatik Corporation EKU07628
Encompass_Preferred
Dog Thrombomodulin (TM) ELISA Kit for detecting TM in plasma. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5W7P8. Gene ID: 474355. Target Synonyms: CD141; THBD; BDCA3; THRM; Fetomodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2928