Learn More
Biomatik Corporation Rat Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07734
Encompass_Preferred
Rat Tissue Inhibitors Of Metalloproteinase 4 (TIMP4) ELISA Kit for detecting TIMP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81556. Gene ID: 680130. Target Synonyms: TIMP Metallopeptidase Inhibitor 4; Tissue Inhibitor of Metalloproteinases 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2946