Learn More
Biomatik Corporation Mouse Toll Like Receptor 7 (TLR7) ELISA Kit
Supplier:  Biomatik Corporation EKU07760
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P58681. Target Synonyms: CD287. Restriction: For Research Use Only.
Catalog No. 50-229-2950