Learn More
Biomatik Corporation Rat Protein Kinase C Alpha (PKCa) ELISA Kit
Supplier:  Biomatik Corporation EKU06916
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.117ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PRKCA; PKC-A; AAG6; PKC-Alpha. Restriction: For Research Use Only.
Catalog No. 50-229-2809