Learn More
Biomatik Corporation Human Protein Kinase, AMP Activated Beta 1 (PRKAb1) ELISA Kit
Supplier:  Biomatik Corporation EKU06930
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.124ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9Y478. Target Synonyms: AMPK; HAMPKb; 5'-AMP-Activated Protein Kinase Subunit Beta-1 Non-Catalytic Subunit; Adenosine Monophosphate Activated Protein Kinase(AMPK). Restriction: For Research Use Only.
Catalog No. 50-229-2811