Learn More
Biomatik Corporation Human Protein Phosphatase 1, Regulatory Subunit 1B (PPP1R1B) ELISA Kit
Supplier:  Biomatik Corporation EKU06937
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9UD71. Target Synonyms: DARPP-32; Dopamine And cAMP Regulated Phosphoprotein; Dopamine and cAMP-regulated neuronal phosphoprotein. Restriction: For Research Use Only.
Catalog No. 50-229-2813