Learn More
Biomatik Corporation Rat Proteinase 3 (PR3) ELISA Kit
Supplier:  Biomatik Corporation EKU06960
Encompass_Preferred
Rat Proteinase 3 (PR3) ELISA Kit for detecting PR3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K597. Gene ID: 314615. Target Synonyms: MBN; PRTN3; ACPA; AGP7; C-ANCA; MBT; P29; PRK; Myeloblastin; Serine Proteinase,Neutrophil,Wegener Granulomatosis Autoantigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2817