Learn More
Biomatik Corporation Rat Pyruvate dehydrogenase alpha 1 (PDHA1) ELISA Kit
Supplier:  Biomatik Corporation EKU06987
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.78-50ng/mL. Sensitivity: 0.29ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PDHCE1A; PHE1A; PHE1-A; PDHa; PDH-A; Pyruvate Dehydrogenase(lipoamide)Alpha 1; Pyruvate Dehydrogenase E1 Component Subunit Alpha,Aomatic Form,Mitochondrial; Pyruvate Dehydrogenase Alpha. Restriction: For Research Use Only.
Catalog No. 50-229-2822