Learn More
Biomatik Corporation Human Reactive Oxygen Species Modulator 1 (ROMO1) ELISA Kit
Supplier:  Biomatik Corporation EKU07016
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: C20orf52; MTGMP; Glyrichin; Mitochondrial Targeting GXXXG Protein; Epididymis tissue protein Li 175; Protein MGR2 homolog. Restriction: For Research Use Only.
Catalog No. 50-229-2828