Learn More
Biomatik Corporation Mouse Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit
Supplier:  Biomatik Corporation EKU07039
Encompass_Preferred
Mouse Regenerating Islet Derived Protein 1 Alpha (REG1a) ELISA Kit for detecting REG1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43137. Gene ID: 19692. Target Synonyms: REG; PSP; ICRF; PSPS1; PTP; Islet cells regeneration factor; Islet of Langerhans regenerating protein; Lithostathine 1 Alpha; Pancreatic Stone Protein; Pancreatic Thread Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2834