Learn More
Biomatik Corporation Rat Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) ELISA Kit
Supplier:  Biomatik Corporation EKU07056
Encompass_Preferred
Rat Regulated On Activation In Normal T-Cell Expressed And Secreted (RANTES) ELISA Kit for detecting RANTES in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50231. Target Synonyms: CCL5; SCYA5; SISd; EoCP; Chemokine C-C-Motif Ligand 5; Eosinophil Chemotactic Cytokine; T-cell Specific Protein p288; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2839