Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein A10 (S100A10) ELISA Kit
Supplier:  Biomatik Corporation EKU07159
Encompass_Preferred
Mouse S100 Calcium Binding Protein A10 (S100A10) ELISA Kit for detecting S100A10 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08207. Gene ID: 20194. Target Synonyms: ANX2L; ANX2LG; CAL1L; CLP11; GP11; P11; P10; Calpactin-1 light chain; Cellular ligand of annexin II; Annexin II Ligand,Calpactin I,Light Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2852