Learn More
Biomatik Corporation Mouse S100 Calcium Binding Protein A11 (S100A11) ELISA Kit
Supplier:  Biomatik Corporation EKU07162
Encompass_Preferred
Mouse S100 Calcium Binding Protein A11 (S100A11) ELISA Kit for detecting S100A11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50543. Gene ID: 20195. Target Synonyms: S100-A11; S100C; MLN70; Protein S100-C; Calgizzarin; Metastatic lymph node gene 70 protein; Protein S100-A11, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2854