Learn More
Biomatik Corporation Human Scavenger Receptor Class A Member 5 (SCARA5) ELISA Kit
Supplier:  Biomatik Corporation EKU07198
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q6ZMJ2. Target Synonyms: NET33; Scavenger receptor hlg. Restriction: For Research Use Only.
Catalog No. 50-229-2858